accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2425-01-6

IUPAC Name: 2-({4-[(oxiran-2-yl)methoxy]phenoxy}methyl)oxirane
Synonyms: 2-[4-(oxiran-2-ylmethoxy)phenoxymethyl]oxirane
Molecular Weight (g/mol): 222.24
Molecular Formula: C12H14O4
InChi Key: FSYPIGPPWAJCJG-UHFFFAOYNA-N
SMILES: C(OC1=CC=C(OCC2CO2)C=C1)C1CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results