accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2425-54-9

IUPAC Name: 1-chlorotetradecane
Synonyms: 1-chlorotetradecane
Molecular Weight (g/mol): 232.84
Molecular Formula: C14H29Cl
InChi Key: RNHWYOLIEJIAMV-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCl
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)