accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2425-77-6

IUPAC Name: 2-hexyldecan-1-ol
Synonyms: hexyldecanol
Molecular Weight (g/mol): 242.45
Molecular Formula: C16H34O
InChi Key: XULHFMYCBKQGEE-UHFFFAOYNA-N
SMILES: CCCCCCCCC(CO)CCCCCC
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)