accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2425-95-8

IUPAC Name: 4-[5-(4-aminophenyl)-1,3,4-oxadiazol-2-yl]aniline
Synonyms: 4-[5-(4-aminophenyl)-1,3,4-oxadiazol-2-yl]aniline
Molecular Weight (g/mol): 252.28
Molecular Formula: C14H12N4O
InChi Key: MJZXFMSIHMJQBW-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=NN=C(O1)C1=CC=C(N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results