accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2(S)IOHO

CAS RN 24250-85-9

IUPAC Name: (2S)-2-amino-3-(4-iodophenyl)propanoic acid
Synonyms: iodo-phenylalanine
Molecular Weight (g/mol): 291.09
Molecular Formula: C9H10INO2
InChi Key: PZNQZSRPDOEBMS-QMMMGPOBSA-N
SMILES: N[C@@H](CC1=CC=C(I)C=C1)C(O)=O
Quantity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)