CAS RN 24253-43-8
IUPAC Name:
4'-bromo-2-(4-bromophenyl)-1,1'-biphenyl
Synonyms:
4'-bromo-2-(4-bromophenyl)-1,1'-biphenyl
Molecular Weight (g/mol):
388.1
Molecular Formula:
C18H12Br2
InChi Key:
CJNMHWJZCGSVQG-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(C=C1)C1=CC=CC=C1C1=CC=C(Br)C=C1
1
–
1
of
1
results
1
4,4″-Dibromo-1,1':2',1″-terphenyl 98.0+%, TCI America™
CAS: 24253-43-8 Molecular Formula: C18H12Br2 Molecular Weight (g/mol): 388.102 MDL Number: MFCD28138092 InChI Key: CJNMHWJZCGSVQG-UHFFFAOYSA-N Synonym: 4,4′C′C-Dibromo-o-terphenyl PubChem CID: 12715106 IUPAC Name: 1,2-bis(4-bromophenyl)benzene SMILES: C1=CC=C(C(=C1)C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br