accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24255-97-8

IUPAC Name: 2-bromo-6-(piperidin-1-yl)pyridine
Synonyms: 2-bromo-6-(piperidin-1-yl)pyridine
Molecular Weight (g/mol): 241.13
Molecular Formula: C10H13BrN2
InChi Key: YHQZMCNKHWPFBS-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=N1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results