accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOCH3O

CAS RN 24262-66-6

IUPAC Name: methyl 4-(acetyloxy)benzoate
Synonyms: methyl 4-(acetyloxy)benzoate
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: PGJQPLVEUVHSFQ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(OC(C)=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results