accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ClClFFF

CAS RN 24279-39-8

IUPAC Name: 2,6-dichloro-4-(trifluoromethyl)aniline
Synonyms: 2,6-dichloro-4-(trifluoromethyl)aniline
Molecular Weight (g/mol): 230.01
Molecular Formula: C7H4Cl2F3N
InChi Key: ITNMAZSPBLRJLU-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(C=C1Cl)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results