accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24295-03-2

IUPAC Name: 1-(1,3-thiazol-2-yl)ethan-1-one
Synonyms: methyl thiazolyl ketone
Molecular Weight (g/mol): 127.16
Molecular Formula: C5H5NOS
InChi Key: MOMFXATYAINJML-UHFFFAOYSA-N
SMILES: CC(=O)C1=NC=CS1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)