accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOON

CAS RN 24297-59-4

IUPAC Name: 2-(1H-indol-1-yl)acetic acid
Synonyms: indol-1-ylacetic acid
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: WQJFIWXYPKYBTO-UHFFFAOYSA-N
SMILES: OC(=O)CN1C=CC2=CC=CC=C12
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)