accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO

CAS RN 2430-16-2

IUPAC Name: 6-phenylhexan-1-ol
Synonyms: 6-phenylhexan-1-ol
Molecular Weight (g/mol): 178.28
Molecular Formula: C12H18O
InChi Key: FDXBUMXUJRZANT-UHFFFAOYSA-N
SMILES: OCCCCCCC1=CC=CC=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)