accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24304-00-5

IUPAC Name: alumanylidyneamine
Synonyms: aluminum nitride aluminium nitride
Molecular Weight (g/mol): 40.99
Molecular Formula: AlN
InChi Key: PIGFYZPCRLYGLF-UHFFFAOYSA-N
SMILES: N#[Al]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (4)
  • (4)
Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (10)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (4)
  • (2)
  • (2)