accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHFNOO

CAS RN 243128-46-3

IUPAC Name: 1-(4-fluoro-2-nitrophenyl)piperazine
Synonyms: 1-(4-fluoro-2-nitrophenyl)piperazine
Molecular Weight (g/mol): 225.22
Molecular Formula: C10H12FN3O2
InChi Key: ZPYLKZRKXDCEQI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(C=CC(F)=C1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results