accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24313-88-0

IUPAC Name: 3,4,5-trimethoxyaniline
Synonyms: 3,4,5-trimethoxyaniline
Molecular Weight (g/mol): 183.21
Molecular Formula: C9H13NO3
InChi Key: XEFRNCLPPFDWAC-UHFFFAOYSA-N
SMILES: COC1=CC(N)=CC(OC)=C1OC
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (2)
Quantity 
  • (3)
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (4)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (5)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)