accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH

CAS RN 2432-11-3

IUPAC Name: 3-phenyl-[1,1'-biphenyl]-2-ol
Synonyms: 3-phenyl-[1,1'-biphenyl]-2-ol
Molecular Weight (g/mol): 246.31
Molecular Formula: C18H14O
InChi Key: ATGFTMUSEPZNJD-UHFFFAOYSA-N
SMILES: OC1=C(C=CC=C1C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results