accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClOHCl

CAS RN 2432-12-4

IUPAC Name: 2,6-dichloro-4-methylphenol
Synonyms: 4-methyl-2,6-dichlorophenol
Molecular Weight (g/mol): 177.02
Molecular Formula: C7H6Cl2O
InChi Key: YXEOEPYIBGTLML-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=C(O)C(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results