accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONOO

CAS RN 2436-96-6

IUPAC Name: 2,2'-dinitro-1,1'-biphenyl
Synonyms: 2,2'-dinitro-biphenyl
Molecular Weight (g/mol): 244.21
Molecular Formula: C12H8N2O4
InChi Key: QAFJHDNFUMKVIE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1C1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)