accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 243666-11-7

IUPAC Name: 7-(trifluoromethyl)quinolin-4-amine
Synonyms: 7-(trifluoromethyl)quinolin-4-amine
Molecular Weight (g/mol): 212.18
Molecular Formula: C10H7F3N2
InChi Key: QZOOIAOULGJTJI-UHFFFAOYSA-N
SMILES: NC1=CC=NC2=CC(=CC=C12)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results