CAS RN 2437-17-4
IUPAC Name:
6-hydroxynaphthalene-1-carboxylic acid
Synonyms:
6-hydroxynaphthalene-1-carboxylic acid
Molecular Weight (g/mol):
188.18
Molecular Formula:
C11H8O3
InChi Key:
JCJUKCIXTRWAQY-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C2C=CC(O)=CC2=CC=C1