CAS RN 24372-48-3
CAS RN 24372-48-3
IUPAC Name:
5-(piperidin-1-yl)thiophene-2-carbaldehyde
Synonyms:
5-(piperidin-1-yl)thiophene-2-carbaldehyde
Molecular Weight (g/mol):
195.28
Molecular Formula:
C10H13NOS
InChi Key:
SFDXRMCPPLZJBG-UHFFFAOYSA-N
SMILES:
O=CC1=CC=C(S1)N1CCCCC1
1
–
1
of
1
results
1
Thermo Scientific Chemicals 5-Piperidinothiophene-2-carbaldehyde, 97%, Thermo Scientific™
CAS: 24372-48-3 Molecular Formula: C10H13NOS Molecular Weight (g/mol): 195.28 MDL Number: MFCD05263177 InChI Key: SFDXRMCPPLZJBG-UHFFFAOYSA-N PubChem CID: 2113341 SMILES: O=CC1=CC=C(S1)N1CCCCC1