accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO

CAS RN 2438-05-3

IUPAC Name: 4-propylbenzoate
Synonyms: 4-propylbenzoate
Molecular Weight (g/mol): 163.2
Molecular Formula: C10H11O2
InChi Key: ATZHGRNFEFVDDJ-UHFFFAOYSA-M
SMILES: CCCC1=CC=C(C=C1)C([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)