accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3(S)ClNOHOOHO(Z)

CAS RN 2438-32-6

IUPAC Name: (2Z)-but-2-enedioic acid; [(3S)-3-(4-chlorophenyl)-3-(pyridin-2-yl)propyl]dimethylamine
Synonyms: (+)-chlorpheniramine; maleic acid
Molecular Weight (g/mol): 390.86
Molecular Formula: C20H23ClN2O4
InChi Key: DBAKFASWICGISY-DASCVMRKSA-N
SMILES: OC(=O)\C=C/C(O)=O.CN(C)CC[C@@H](C1=CC=C(Cl)C=C1)C1=CC=CC=N1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)