accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2439-04-5

IUPAC Name: isoquinolin-5-ol
Synonyms: isoquinolin-5-ol
Molecular Weight (g/mol): 145.16
Molecular Formula: C9H7NO
InChi Key: CSNXUYRHPXGSJD-UHFFFAOYSA-N
SMILES: OC1=C2C=CN=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results