accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2439-55-6

H3CNHCH3

CAS RN 2439-55-6

IUPAC Name: methyl(octadecyl)amine
Synonyms: 1-octadecanamine, N-methyl-
Molecular Weight (g/mol): 283.54
Molecular Formula: C19H41N
InChi Key: SZEGKVHRCLBFKJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCNC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)