accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2439-85-2

IUPAC Name: 2-bromo-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-bromoisoindole-1,3-dione
Molecular Weight (g/mol): 226.03
Molecular Formula: C8H4BrNO2
InChi Key: MARXMDRWROUXMD-UHFFFAOYSA-N
SMILES: BrN1C(=O)C2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)