accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOCH3HOOH

CAS RN 243990-53-6

IUPAC Name: [4-(benzyloxy)-3-methoxyphenyl]boronic acid
Synonyms: 4-(benzyloxy)-3-methoxyphenylboronic acid
Molecular Weight (g/mol): 258.08
Molecular Formula: C14H15BO4
InChi Key: PBVVDNNISDIUSU-UHFFFAOYSA-N
SMILES: COC1=C(OCC2=CC=CC=C2)C=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results