accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3SiCH2CH3H3CCH3H3C

CAS RN 24400-84-8

IUPAC Name: (prop-2-en-1-yl)tris(propan-2-yl)silane
Synonyms: triisopropyl(prop-2-en-1-yl)silane
Molecular Weight (g/mol): 198.43
Molecular Formula: C12H26Si
InChi Key: AKQHUJRZKBYZLC-UHFFFAOYSA-N
SMILES: CC(C)[Si](CC=C)(C(C)C)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results