accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFH3CNNCH3

CAS RN 244193-50-8

IUPAC Name: 1-hexyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
Synonyms: 1-hexyl-3-methylimidazolium tetrafluoroborate
Molecular Weight (g/mol): 254.08
Molecular Formula: C10H19BF4N2
InChi Key: MFXLOVLEQJRXFP-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CCCCCCN1C=C[N+](C)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results