accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFH3CNNCH3

CAS RN 244193-52-0

IUPAC Name: 3-methyl-1-octyl-1H-imidazol-3-ium; tetrafluoroboranuide
Synonyms: 1-octyl-3-methylimidazolium tetrafluoroborate
Molecular Weight (g/mol): 282.13
Molecular Formula: C12H23BF4N2
InChi Key: GXZCAMSPWNHTAE-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CCCCCCCCN1C=C[N+](C)=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)