accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 2442-49-1

IUPAC Name: methyl tetracosanoate
Synonyms: methyl tetracosanoate
Molecular Weight (g/mol): 382.67
Molecular Formula: C25H50O2
InChi Key: XUDJZDNUVZHSKZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCC(=O)OC
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)