accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBOHBr

CAS RN 244205-40-1

IUPAC Name: (2-bromophenyl)boronic acid
Synonyms: 2-bromophenylboronic acid
Molecular Weight (g/mol): 200.83
Molecular Formula: C6H6BBrO2
InChi Key: PLVCYMZAEQRYHJ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=C1Br
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (5)
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (3)