accessibility menu, dialog, popup

UserName

Viewing profile as user:
NClI

CAS RN 244221-57-6

IUPAC Name: 5-chloro-2-iodopyridine
Synonyms: 5-chloro-2-iodopyridine
Molecular Weight (g/mol): 239.44
Molecular Formula: C5H3ClIN
InChi Key: CXWLXKZIXLOBCC-UHFFFAOYSA-N
SMILES: ClC1=CN=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results