accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24425-40-9

IUPAC Name: 2,3-dihydro-1H-inden-5-amine
Synonyms: indan-5-ylamine
Molecular Weight (g/mol): 133.19
Molecular Formula: C9H11N
InChi Key: LEWZOBYWGWKNCK-UHFFFAOYSA-N
SMILES: NC1=CC=C2CCCC2=C1
Boiling Point 
  • (2)
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)