accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOPOOP(?)

CAS RN 244261-66-3

IUPAC Name: {4-[5-(diphenylphosphanyl)-2H-1,3-benzodioxol-4-yl]-2H-1,3-benzodioxol-5-yl}diphenylphosphane
Synonyms: segphos
Molecular Weight (g/mol): 610.59
Molecular Formula: C38H28O4P2
InChi Key: RZZDRSHFIVOQAF-UHFFFAOYSA-N
SMILES: C1OC2=C(O1)C(=C(C=C2)P(C1=CC=CC=C1)C1=CC=CC=C1)C1=C(C=CC2=C1OCO2)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)