accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOCl

CAS RN 2444-36-2

IUPAC Name: 2-(2-chlorophenyl)acetic acid
Synonyms: (2-chlorophenyl)acetic acid
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: IUJAAIZKRJJZGQ-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)