CAS RN 24443-15-0
IUPAC Name:
1,3-dibromo-3-methylbutane
Synonyms:
1,3-dibromo-3-methylbutane
Molecular Weight (g/mol):
229.94
Molecular Formula:
C5H10Br2
InChi Key:
XMJFMQSXQQUJTQ-UHFFFAOYSA-N
SMILES:
CC(C)(Br)CCBr
1
–
1
of
1
results
1
Promotions Available
1,3-Dibromo-3-methylbutane, 98%
1,3-Dibromo-3-methylbutane, 98%
CAS: 24443-15-0 Molecular Formula: C5H10Br2 Molecular Weight (g/mol): 229.94 MDL Number: MFCD00060773 InChI Key: XMJFMQSXQQUJTQ-UHFFFAOYSA-N Synonym: butane, 1,3-dibromo-3-methyl,acmc-20ap4g,2,4-dibromo-2-methylbutane PubChem CID: 141113 IUPAC Name: 1,3-dibromo-3-methylbutane SMILES: CC(C)(Br)CCBr