accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONHClCl

CAS RN 24448-73-5

IUPAC Name: N-(3,4-dichlorophenyl)-2-hydroxybenzamide
Synonyms: N-(3,4-dichlorophenyl)-2-hydroxybenzamide
Molecular Weight (g/mol): 282.12
Molecular Formula: C13H9Cl2NO2
InChi Key: XWZCOIHEGFREJR-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C(=O)NC1=CC=C(Cl)C(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results