accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2445-83-2

IUPAC Name: 7-methyl-2H-chromen-2-one
Synonyms: 7-methylchromen-2-one
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: DLHXRDUXNVEIEY-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC(=O)OC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results