CAS RN 244761-29-3
IUPAC Name:
lithium(1+) tetraoxo-1,4,6,9-tetraoxa-5-boraspiro[4.4]nonan-5-uide
Synonyms:
lithium(1+) tetraoxo-1,4,6,9-tetraoxa-5-boraspiro[4.4]nonan-5-uide
Molecular Weight (g/mol):
193.79
Molecular Formula:
C4BLiO8
InChi Key:
NVQAYVUCVASGDK-UHFFFAOYSA-N
SMILES:
[Li+].O=C1O[B-]2(OC1=O)OC(=O)C(=O)O2
1
–
1
of
1
results
1
Sigma Aldrich Lithium bis(oxalato)borate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.