accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2448-06-8

IUPAC Name: 3-nitro-N,N-bis(propan-2-yl)benzamide
Synonyms: N,N-diisopropyl-3-nitrobenzamide
Molecular Weight (g/mol): 250.3
Molecular Formula: C13H18N2O3
InChi Key: UFFKMMNKRCCXPE-UHFFFAOYSA-N
SMILES: CC(C)N(C(C)C)C(=O)C1=CC=CC(=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results