accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24484-22-8

OOF

CAS RN 24484-22-8

IUPAC Name: 5-(4-fluorophenyl)pentanoate
Synonyms: 5-(4-fluorophenyl)pentanoate
Molecular Weight (g/mol): 195.21
Molecular Formula: C11H12FO2
InChi Key: BXEFPLJKYWEEAN-UHFFFAOYSA-M
SMILES: [O-]C(=O)CCCCC1=CC=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results