accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2450-71-7

IUPAC Name: prop-2-yn-1-aminium chloride
Synonyms: prop-2-yn-1-aminium chloride
Molecular Weight (g/mol): 91.54
Molecular Formula: C3H6ClN
InChi Key: IKXNIQJDNKPPCH-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]CC#C
Boiling Point 
  • (2)
  • (4)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (6)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (6)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (6)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (6)