accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOHOONOOH

CAS RN 245049-70-1

IUPAC Name: 4,9-dihydroxy-4,9-diazatricyclo[5.3.0.0²,⁶]decane-3,5,8,10-tetrone
Synonyms: 4,9-dihydroxy-4,9-diazatricyclo[5.3.0.0²,⁶]decane-3,5,8,10-tetrone
Molecular Weight (g/mol): 226.14
Molecular Formula: C8H6N2O6
InChi Key: HONIHRUONDEYHT-UHFFFAOYSA-N
SMILES: ON1C(=O)C2C(C3C2C(=O)N(O)C3=O)C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results