accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24521-76-4

ONHSNHNHN

CAS RN 24521-76-4

IUPAC Name: 6-sulfanylidene-1H,2H,4H,5H,6H-pyrazolo[3,4-d]pyrimidin-4-one
Synonyms: 6-sulfanylidene-1H,2H,5H-pyrazolo[3,4-d]pyrimidin-4-one
Molecular Weight (g/mol): 168.17
Molecular Formula: C5H4N4OS
InChi Key: SXRSXYWROQWSGJ-UHFFFAOYSA-N
SMILES: O=C1NC(=S)N=C2NNC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results