accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2455-24-5

IUPAC Name: (oxolan-2-yl)methyl 2-methylprop-2-enoate
Synonyms: tetrahydrofurfuryl methacrylate
Molecular Weight (g/mol): 170.21
Molecular Formula: C9H14O3
InChi Key: LCXXNKZQVOXMEH-UHFFFAOYNA-N
SMILES: CC(=C)C(=O)OCC1CCCO1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)