accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOO

CAS RN 24552-27-0

IUPAC Name: phenyl 3-chloropropanoate
Synonyms: phenyl 3-chloropropanoate
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: RAFRTSDUWORDLA-UHFFFAOYSA-N
SMILES: ClCCC(=O)OC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results