accessibility menu, dialog, popup

UserName

Viewing profile as user:
SrH3CCH3OOH3CCH3OO

CAS RN 2457-02-5

IUPAC Name: strontium(2+) bis(2-ethylhexanoate)
Synonyms: strontium(2+) bis(2-ethylhexanoate)
Molecular Weight (g/mol): 374.03
Molecular Formula: C16H30O4Sr
InChi Key: DLWBHRIWCMOQKI-UHFFFAOYNA-L
SMILES: [Sr++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)