accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 2459-25-8

IUPAC Name: methyl naphthalene-2-carboxylate
Synonyms: methyl naphthalene-2-carboxylate
Molecular Weight (g/mol): 186.21
Molecular Formula: C12H10O2
InChi Key: IODOXLXFXNATGI-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C2C=CC=CC2=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)