accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrH3CH2N

CAS RN 24596-19-8

IUPAC Name: 4-bromo-2,6-dimethylaniline
Synonyms: 4-bromo-2,6-dimethylaniline
Molecular Weight (g/mol): 200.08
Molecular Formula: C8H10BrN
InChi Key: QGLAYJCJLHNIGJ-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC(C)=C1N
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (7)
  • (2)
  • (8)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)